Product Details

SNP ID
rs200767481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28908718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCCTTGGTTTTCCTCATTCAAC[C/T]GCTTTACCTCATGGTGCAGGCACTT
Phenotype
MIM: 108730 MIM: 611869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP2A1 PubMed Links

Gene Details

Gene
ATP2A1
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1
There are no transcripts associated with this gene.

Gene
RABEP2
Gene Name
rabaptin, RAB GTPase binding effector protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024816.2 1184 Missense Mutation NP_079092.2

View Full Product Details