Product Details

SNP ID
rs201499017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30555643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCGGCGGCCACAGTCGGCGCAGC[C/G]ATAGGGTTTCTCGCCGGTGTGGACG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF764 PubMed Links

Gene Details

Gene
ZNF764
Gene Name
zinc finger protein 764
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172679.1 917 Missense Mutation CGC,GGC R259G NP_001166150.1
NM_033410.3 917 Missense Mutation CGC,GGC R260G NP_219363.2

View Full Product Details