Product Details

SNP ID
rs199530258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89722813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGGTCAACGCCTCCCCGGCTG[A/G]TCGCCGGAAGCCTTGTGCAAAGTAC
Phenotype
MIM: 608460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS9D1 PubMed Links

Gene Details

Gene
VPS9D1
Gene Name
VPS9 domain containing 1
There are no transcripts associated with this gene.

Gene
VPS9D1-AS1
Gene Name
VPS9D1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNF276
Gene Name
zinc finger protein 276
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113525.1 575 Missense Mutation GAT,GGT D163G NP_001106997.1
NM_152287.3 575 Missense Mutation GAT,GGT D88G NP_689500.2
XM_005256324.3 575 Missense Mutation GAT,GGT D163G XP_005256381.1
XM_005256328.3 575 Intron XP_005256385.1
XM_017023889.1 575 Missense Mutation GAT,GGT D163G XP_016879378.1
XM_017023890.1 575 UTR 5 XP_016879379.1

View Full Product Details