Product Details

SNP ID
rs200450867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30408181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTGGTGAAACTCAAGATCCCCA[G/T]GGACAACAAGGAGGTATGTGTCACA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF48 PubMed Links

Gene Details

Gene
ZNF48
Gene Name
zinc finger protein 48
There are no transcripts associated with this gene.

Gene
ZNF771
Gene Name
zinc finger protein 771
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142305.1 401 Missense Mutation AGG,ATG R43M NP_001135777.1
NM_016643.3 401 Missense Mutation AGG,ATG R43M NP_057727.2

View Full Product Details