Product Details

SNP ID
rs200748731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28932939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTTCCGGTGGAATGTTTCGGA[C/T]CTAGGTGGCCTGGGCTGTGGCCTGA
Phenotype
MIM: 107265 MIM: 611869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD19 PubMed Links

Gene Details

Gene
CD19
Gene Name
CD19 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178098.1 247 Silent Mutation GAC,GAT D128D NP_001171569.1
NM_001770.5 247 Silent Mutation GAC,GAT D128D NP_001761.3
XM_006721103.3 247 Silent Mutation GAC,GAT D39D XP_006721166.1
XM_011545981.1 247 Silent Mutation GAC,GAT D128D XP_011544283.1
XM_017023893.1 247 Silent Mutation GAC,GAT D39D XP_016879382.1
Gene
RABEP2
Gene Name
rabaptin, RAB GTPase binding effector protein 2
There are no transcripts associated with this gene.

View Full Product Details