Product Details

SNP ID
rs201232409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67661045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCCGCCCGGCCCCGCCATGGCC[C/G]GGCCGCAGAGGACTCCGGCGCGCAG
Phenotype
MIM: 609377 MIM: 610859 MIM: 607484
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACD PubMed Links

Gene Details

Gene
ACD
Gene Name
adrenocortical dysplasia homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082486.1 81 Intron NP_001075955.1
NM_001082487.1 81 Intron NP_001075956.1
NM_022914.2 81 Intron NP_075065.2
XM_005256115.3 81 Intron XP_005256172.1
Gene
C16orf86
Gene Name
chromosome 16 open reading frame 86
There are no transcripts associated with this gene.

Gene
CARMIL2
Gene Name
capping protein regulator and myosin 1 linker 2
There are no transcripts associated with this gene.

Gene
ENKD1
Gene Name
enkurin domain containing 1
There are no transcripts associated with this gene.

Gene
PARD6A
Gene Name
par-6 family cell polarity regulator alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037281.1 81 Missense Mutation CGG,GGG R3G NP_001032358.1
NM_016948.2 81 Missense Mutation CGG,GGG R3G NP_058644.1
XM_005255977.3 81 Missense Mutation CGG,GGG R3G XP_005256034.1
XM_011523095.2 81 Missense Mutation CGG,GGG R3G XP_011521397.1
XM_011523096.2 81 Missense Mutation CGG,GGG R3G XP_011521398.1
XM_017023261.1 81 Missense Mutation CGG,GGG R3G XP_016878750.1

View Full Product Details