Product Details
- SNP ID
-
rs199867890
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:723930 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGATGAGAGCAGTGCCCCAGGCC[C/T]GCCGTGCTACAGAGGTTTGTGGTGG
- Phenotype
-
MIM: 614666
MIM: 610998
MIM: 611118
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC78
PubMed Links
Gene Details
- Gene
- CCDC78
- Gene Name
- coiled-coil domain containing 78
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001031737.2 |
1361 |
Missense Mutation |
AGG,GGG |
R354G |
NP_001026907.2 |
XM_006720838.1 |
1361 |
Missense Mutation |
AGG,GGG |
R428G |
XP_006720901.1 |
XM_006720843.3 |
1361 |
Missense Mutation |
AGG,GGG |
R354G |
XP_006720906.1 |
XM_011522356.1 |
1361 |
Missense Mutation |
AGG,GGG |
R503G |
XP_011520658.1 |
XM_011522357.1 |
1361 |
Missense Mutation |
AGG,GGG |
R499G |
XP_011520659.1 |
XM_011522358.2 |
1361 |
Missense Mutation |
AGG,GGG |
R503G |
XP_011520660.1 |
XM_011522359.1 |
1361 |
Missense Mutation |
AGG,GGG |
R492G |
XP_011520661.1 |
XM_011522360.1 |
1361 |
Missense Mutation |
AGG,GGG |
R488G |
XP_011520662.1 |
XM_011522361.1 |
1361 |
Missense Mutation |
AGG,GGG |
R503G |
XP_011520663.1 |
XM_011522362.1 |
1361 |
Missense Mutation |
AGG,GGG |
R503G |
XP_011520664.1 |
XM_011522363.1 |
1361 |
Missense Mutation |
AGG,GGG |
R503G |
XP_011520665.1 |
XM_011522364.1 |
1361 |
Missense Mutation |
AGG,GGG |
R503G |
XP_011520666.1 |
XM_011522365.1 |
1361 |
Missense Mutation |
AGG,GGG |
R432G |
XP_011520667.1 |
XM_011522366.1 |
1361 |
Missense Mutation |
AGG,GGG |
R429G |
XP_011520668.1 |
XM_011522367.1 |
1361 |
Missense Mutation |
AGG,GGG |
R376G |
XP_011520669.1 |
XM_011522368.1 |
1361 |
Missense Mutation |
AGG,GGG |
R372G |
XP_011520670.1 |
XM_011522369.1 |
1361 |
Missense Mutation |
AGG,GGG |
R358G |
XP_011520671.1 |
XM_011522370.1 |
1361 |
Missense Mutation |
AGG,GGG |
R302G |
XP_011520672.1 |
XM_011522371.2 |
1361 |
Missense Mutation |
AGG,GGG |
R207G |
XP_011520673.1 |
XM_017022929.1 |
1361 |
Missense Mutation |
AGG,GGG |
R503G |
XP_016878418.1 |
XM_017022930.1 |
1361 |
Missense Mutation |
AGG,GGG |
R203G |
XP_016878419.1 |
XM_017022931.1 |
1361 |
UTR 5 |
|
|
XP_016878420.1 |
XM_017022932.1 |
1361 |
Missense Mutation |
AGG,GGG |
R113G |
XP_016878421.1 |
- Gene
- FAM173A
- Gene Name
- family with sequence similarity 173 member A
There are no transcripts associated with this gene.
- Gene
- HAGHL
- Gene Name
- hydroxyacylglutathione hydrolase-like
There are no transcripts associated with this gene.
- Gene
- METRN
- Gene Name
- meteorin, glial cell differentiation regulator
There are no transcripts associated with this gene.
- Gene
- NARFL
- Gene Name
- nuclear prelamin A recognition factor like
There are no transcripts associated with this gene.
View Full Product Details