Product Details

SNP ID
rs201755210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88646192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGCAGGAAGCCGGCGGGCACC[A/G]AGAGCCTGGGGGACAGCGGGTGAGA
Phenotype
MIM: 608508 MIM: 604628 MIM: 603236
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYBA PubMed Links

Gene Details

Gene
CYBA
Gene Name
cytochrome b-245 alpha chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000101.3 364 Missense Mutation NP_000092.2
XM_011522905.2 364 Missense Mutation XP_011521207.1
Gene
IL17C
Gene Name
interleukin 17C
There are no transcripts associated with this gene.

Gene
MVD
Gene Name
mevalonate diphosphate decarboxylase
There are no transcripts associated with this gene.

View Full Product Details