Product Details

SNP ID
rs201951347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3029379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGGGCTCTAAGATCTCTTCCGG[A/G]GGTGAAGGCTGGGGGCTGGACGCCT
Phenotype
MIM: 617003 MIM: 615403 MIM: 605914
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BICDL2 PubMed Links

Gene Details

Gene
BICDL2
Gene Name
BICD family like cargo adaptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103175.1 1114 Silent Mutation CCC,CCT P336P NP_001096645.1
XM_005255135.4 1114 Silent Mutation CCC,CCT P336P XP_005255192.1
XM_011522391.1 1114 Silent Mutation CCC,CCT P271P XP_011520693.1
Gene
HCFC1R1
Gene Name
host cell factor C1 regulator 1
There are no transcripts associated with this gene.

Gene
LOC100128770
Gene Name
uncharacterized LOC100128770
There are no transcripts associated with this gene.

Gene
THOC6
Gene Name
THO complex 6
There are no transcripts associated with this gene.

Gene
TNFRSF12A
Gene Name
TNF receptor superfamily member 12A
There are no transcripts associated with this gene.

View Full Product Details