Product Details

SNP ID
rs201890427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2461110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTGCTGTCAGTGGGGGATGGGA[C/G]CCGTGTTGGGATGGGAGCCCGAACC
Phenotype
MIM: 600227
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C16orf59 PubMed Links

Gene Details

Gene
C16orf59
Gene Name
chromosome 16 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025108.2 426 Missense Mutation ACC,AGC T164S NP_079384.2
XM_011522667.1 426 Missense Mutation ACC,AGC T131S XP_011520969.1
Gene
CCNF
Gene Name
cyclin F
There are no transcripts associated with this gene.

Gene
LOC729652
Gene Name
uncharacterized LOC729652
There are no transcripts associated with this gene.

Gene
MIR6768
Gene Name
microRNA 6768
There are no transcripts associated with this gene.

View Full Product Details