Product Details

SNP ID
rs202224384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85099232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGATGCCCACCCTGCCCACACA[C/T]ACAGTGGGCTGGCTGCCCCTTAACC
Phenotype
MIM: 611675
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM92B PubMed Links

Gene Details

Gene
FAM92B
Gene Name
family with sequence similarity 92 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198491.2 1024 Missense Mutation ATG,GTG M290V NP_940893.1
XM_005255931.2 1024 Intron XP_005255988.1
XM_011523063.1 1024 Missense Mutation ATG,GTG M290V XP_011521365.1
XM_017023198.1 1024 Intron XP_016878687.1
Gene
KIAA0513
Gene Name
KIAA0513
There are no transcripts associated with this gene.

View Full Product Details