Product Details

SNP ID
rs201641851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2276692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCCTCATCGCCCCTCCTCTGC[A/G]GTGGGCGGCTGCAGGTGGGCGAAGC
Phenotype
MIM: 601615 MIM: 606447
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA3 PubMed Links

Gene Details

Gene
ABCA3
Gene Name
ATP binding cassette subfamily A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001089.2 5809 Silent Mutation ACC,ACT T1699T NP_001080.2
Gene
LOC106660606
Gene Name
uncharacterized LOC106660606
There are no transcripts associated with this gene.

Gene
MIR3677
Gene Name
microRNA 3677
There are no transcripts associated with this gene.

Gene
MIR4717
Gene Name
microRNA 4717
There are no transcripts associated with this gene.

Gene
MIR940
Gene Name
microRNA 940
There are no transcripts associated with this gene.

Gene
RNPS1
Gene Name
RNA binding protein with serine rich domain 1
There are no transcripts associated with this gene.

View Full Product Details