Product Details

SNP ID
rs202065363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30555349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATGAACCCACTGGTGTTTGGCCA[C/T]GGCTGACTTCTGGCCAAAGCGGCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF764 PubMed Links

Gene Details

Gene
ZNF764
Gene Name
zinc finger protein 764
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172679.1 1211 Missense Mutation ATG,GTG M357V NP_001166150.1
NM_033410.3 1211 Missense Mutation ATG,GTG M358V NP_219363.2

View Full Product Details