Product Details
- SNP ID
-
rs201836260
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:50751613 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTCTTAAAAACTAGGAAGAAGGT[C/T]GTGGTCAAGGTTTCACTGACGGGGT
- Phenotype
-
MIM: 605018
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYLD
PubMed Links
Gene Details
- Gene
- CYLD
- Gene Name
- CYLD lysine 63 deubiquitinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001042355.1 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
NP_001035814.1 |
NM_001042412.1 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
NP_001035877.1 |
NM_015247.2 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
NP_056062.1 |
XM_005255812.2 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
XP_005255869.1 |
XM_006721149.1 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
XP_006721212.1 |
XM_011522907.2 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
XP_011521209.1 |
XM_011522908.1 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
XP_011521210.1 |
XM_011522911.2 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
XP_011521213.1 |
XM_017022977.1 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
XP_016878466.1 |
XM_017022978.1 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
XP_016878467.1 |
XM_017022979.1 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
XP_016878468.1 |
XM_017022980.1 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
XP_016878469.1 |
XM_017022981.1 |
953 |
Missense Mutation |
CGT,TGT |
R172C |
XP_016878470.1 |
- Gene
- MIR3181
- Gene Name
- microRNA 3181
There are no transcripts associated with this gene.
View Full Product Details