Product Details

SNP ID
rs202131705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3023402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGGCAGGCTGGGATACTGACA[C/G]AGGAGGCAGCCTGTTGGGGACCAGA
Phenotype
MIM: 617003 MIM: 615798 MIM: 615799 MIM: 615403 MIM: 605914
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BICDL2 PubMed Links

Gene Details

Gene
BICDL2
Gene Name
BICD family like cargo adaptor 2
There are no transcripts associated with this gene.

Gene
CLDN6
Gene Name
claudin 6
There are no transcripts associated with this gene.

Gene
CLDN9
Gene Name
claudin 9
There are no transcripts associated with this gene.

Gene
HCFC1R1
Gene Name
host cell factor C1 regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002017.2 451 Intron NP_001002017.1
NM_001002018.2 451 Intron NP_001002018.1
NM_001288665.1 451 Intron NP_001275594.1
NM_001288666.1 451 Intron NP_001275595.1
NM_001288667.1 451 Missense Mutation CTG,GTG L36V NP_001275596.1
NM_001288668.1 451 UTR 5 NP_001275597.1
NM_001308070.1 451 Intron NP_001294999.1
NM_017885.3 451 Intron NP_060355.1
XM_011522559.2 451 Intron XP_011520861.1
XM_017023384.1 451 Missense Mutation CTG,GTG L36V XP_016878873.1
Gene
LOC100128770
Gene Name
uncharacterized LOC100128770
There are no transcripts associated with this gene.

Gene
THOC6
Gene Name
THO complex 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142350.1 451 Intron NP_001135822.1
NM_024339.3 451 Intron NP_077315.2
Gene
TNFRSF12A
Gene Name
TNF receptor superfamily member 12A
There are no transcripts associated with this gene.

View Full Product Details