Product Details

SNP ID
rs202034447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67154766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGTGGCGCGGCGGCCCTCGGC[C/G]TGCACGAAGCGCCGCAGCAGCTGGA
Phenotype
MIM: 609077 MIM: 602438 MIM: 603500
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
B3GNT9 PubMed Links

Gene Details

Gene
B3GNT9
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9
There are no transcripts associated with this gene.

Gene
C16orf70
Gene Name
chromosome 16 open reading frame 70
There are no transcripts associated with this gene.

Gene
FBXL8
Gene Name
F-box and leucine rich repeat protein 8
There are no transcripts associated with this gene.

Gene
HSF4
Gene Name
heat shock transcription factor 4
There are no transcripts associated with this gene.

Gene
TRADD
Gene Name
TNFRSF1A associated via death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323552.1 915 Missense Mutation CAC,CAG H274Q NP_001310481.1
NM_003789.3 915 Missense Mutation CAC,CAG H274Q NP_003780.1
XM_005256213.3 915 Missense Mutation CAC,CAG H214Q XP_005256270.1
XM_017023815.1 915 Missense Mutation CAC,CAG H274Q XP_016879304.1

View Full Product Details