Product Details
- SNP ID
-
rs201784585
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:68316035 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCACGGGGTCTGTGGAGTGGCTGG[A/G]GGAGGATGAACACTATGATTACCAC
- Phenotype
-
MIM: 610087
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRMT7
PubMed Links
Gene Details
- Gene
- PRMT7
- Gene Name
- protein arginine methyltransferase 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001184824.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
NP_001171753.1 |
NM_001290018.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
NP_001276947.1 |
NM_019023.2 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
NP_061896.1 |
XM_011523112.2 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_011521414.1 |
XM_011523113.2 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_011521415.1 |
XM_011523115.2 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_011521417.1 |
XM_011523116.2 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_011521418.1 |
XM_011523121.2 |
197 |
UTR 5 |
|
|
XP_011521423.1 |
XM_011523124.2 |
197 |
UTR 5 |
|
|
XP_011521426.1 |
XM_011523125.2 |
197 |
UTR 5 |
|
|
XP_011521427.1 |
XM_011523126.2 |
197 |
UTR 5 |
|
|
XP_011521428.1 |
XM_011523128.2 |
197 |
UTR 5 |
|
|
XP_011521430.1 |
XM_011523131.2 |
197 |
UTR 5 |
|
|
XP_011521433.1 |
XM_017023290.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878779.1 |
XM_017023291.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878780.1 |
XM_017023292.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878781.1 |
XM_017023293.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878782.1 |
XM_017023294.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878783.1 |
XM_017023295.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878784.1 |
XM_017023296.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878785.1 |
XM_017023297.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878786.1 |
XM_017023298.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878787.1 |
XM_017023299.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878788.1 |
XM_017023300.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878789.1 |
XM_017023301.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878790.1 |
XM_017023302.1 |
197 |
UTR 5 |
|
|
XP_016878791.1 |
XM_017023303.1 |
197 |
UTR 5 |
|
|
XP_016878792.1 |
XM_017023304.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878793.1 |
XM_017023305.1 |
197 |
UTR 5 |
|
|
XP_016878794.1 |
XM_017023306.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878795.1 |
XM_017023307.1 |
197 |
UTR 5 |
|
|
XP_016878796.1 |
XM_017023308.1 |
197 |
UTR 5 |
|
|
XP_016878797.1 |
XM_017023309.1 |
197 |
UTR 5 |
|
|
XP_016878798.1 |
XM_017023310.1 |
197 |
UTR 5 |
|
|
XP_016878799.1 |
XM_017023311.1 |
197 |
UTR 5 |
|
|
XP_016878800.1 |
XM_017023312.1 |
197 |
Missense Mutation |
GAG,GGG |
E19G |
XP_016878801.1 |
XM_017023313.1 |
197 |
UTR 5 |
|
|
XP_016878802.1 |
XM_017023314.1 |
197 |
UTR 5 |
|
|
XP_016878803.1 |
XM_017023315.1 |
197 |
UTR 5 |
|
|
XP_016878804.1 |
XM_017023316.1 |
197 |
UTR 5 |
|
|
XP_016878805.1 |
- Gene
- SLC7A6OS
- Gene Name
- solute carrier family 7 member 6 opposite strand
There are no transcripts associated with this gene.
View Full Product Details