Product Details

SNP ID
rs201879579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:20311269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGTCAGGAGGACCATAGGCCAG[A/G]CCACCAGGAACCCTGAAATACAAGA
Phenotype
MIM: 602977
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GP2 PubMed Links

Gene Details

Gene
GP2
Gene Name
glycoprotein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007240.2 1502 Missense Mutation GCC,GTC A523V NP_001007241.2
NM_001007241.2 1502 Missense Mutation GCC,GTC A376V NP_001007242.2
NM_001007242.2 1502 Missense Mutation GCC,GTC A373V NP_001007243.2
NM_001502.3 1502 Missense Mutation GCC,GTC A520V NP_001493.2
XM_017023155.1 1502 Missense Mutation GCC,GTC A501V XP_016878644.1

View Full Product Details