Product Details

SNP ID
rs199833326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42798669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAATCCAGAGGGTCACCAGCTC[C/T]CGAAGACGCCATGTTGCCACTCCTC
Phenotype
MIM: 614775 MIM: 601844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNTD1 PubMed Links

Gene Details

Gene
CNTD1
Gene Name
cyclin N-terminal domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173478.2 57 Intron NP_775749.2
XM_005257043.4 57 UTR 5 XP_005257100.1
XM_005257044.4 57 Intron XP_005257101.1
XM_005257045.4 57 Intron XP_005257102.1
XM_006721690.2 57 Intron XP_006721753.1
XM_011524310.2 57 Intron XP_011522612.1
XM_011524311.2 57 Intron XP_011522613.1
XM_017024171.1 57 Intron XP_016879660.1
Gene
COA3
Gene Name
cytochrome c oxidase assembly factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040431.2 57 Missense Mutation AGA,GGA R5G NP_001035521.1
Gene
WNK4
Gene Name
WNK lysine deficient protein kinase 4
There are no transcripts associated with this gene.

View Full Product Details