Product Details

SNP ID
rs199565053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32488387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGGTGGAGAGCTGCAAGGAGG[C/T]CTTTTGGGACCGTTGCCTCTCTGTC
Phenotype
MIM: 603460 MIM: 606539 MIM: 604449
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK5R1 PubMed Links

Gene Details

Gene
CDK5R1
Gene Name
cyclin dependent kinase 5 regulatory subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003885.2 975 Missense Mutation GCC,GTC A256V NP_003876.1
XM_011525438.2 975 Missense Mutation GCC,GTC A256V XP_011523740.1
XM_017025281.1 975 Missense Mutation GCC,GTC A256V XP_016880770.1
Gene
MYO1D
Gene Name
myosin ID
There are no transcripts associated with this gene.

Gene
PSMD11
Gene Name
proteasome 26S subunit, non-ATPase 11
There are no transcripts associated with this gene.

View Full Product Details