Product Details

SNP ID
rs199801892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43216706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATTTGGTTGACAGATGTTTTAC[A/G]CAGTAGCTACCAAGATACGGGATGA
Phenotype
MIM: 166945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00854 PubMed Links

Gene Details

Gene
LINC00854
Gene Name
long intergenic non-protein coding RNA 854
There are no transcripts associated with this gene.

Gene
NBR1
Gene Name
NBR1, autophagy cargo receptor
There are no transcripts associated with this gene.

Gene
TMEM106A
Gene Name
transmembrane protein 106A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291586.1 865 Missense Mutation ACA,GCA T194A NP_001278515.1
NM_001291587.1 865 Missense Mutation ACA,GCA T146A NP_001278516.1
NM_001291588.1 865 Missense Mutation ACA,GCA T147A NP_001278517.1
NM_145041.3 865 Missense Mutation ACA,GCA T194A NP_659478.1
XM_006721658.2 865 Missense Mutation ACA,GCA T194A XP_006721721.1
XM_006721659.3 865 Missense Mutation ACA,GCA T146A XP_006721722.1
XM_011524260.1 865 Missense Mutation ACA,GCA T147A XP_011522562.1
XM_017024113.1 865 Missense Mutation ACA,GCA T194A XP_016879602.1
XM_017024114.1 865 Missense Mutation ACA,GCA T146A XP_016879603.1

View Full Product Details