Product Details

SNP ID
rs199846495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29574727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTGCTTCTTCTCTCGGGTGGTG[A/G]TGGTGACCAGCTGCTTGGCAGCCTT
Phenotype
MIM: 608434
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD15 PubMed Links

Gene Details

Gene
ABHD15
Gene Name
abhydrolase domain containing 15
There are no transcripts associated with this gene.

Gene
GIT1
Gene Name
GIT ArfGAP 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085454.1 2475 Missense Mutation ACC,ATC T763I NP_001078923.1
NM_014030.3 2475 Missense Mutation ACC,ATC T754I NP_054749.2
XM_011524684.1 2475 Missense Mutation ACC,ATC T770I XP_011522986.1
XM_011524685.1 2475 Missense Mutation ACC,ATC T761I XP_011522987.1
Gene
TP53I13
Gene Name
tumor protein p53 inducible protein 13
There are no transcripts associated with this gene.

View Full Product Details