Product Details

SNP ID
rs200189113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78216689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAACATAAAAAGCATTCGTCC[A/G]GTTGCGCTTTCCTTTCTGTCAAGAA
Phenotype
MIM: 603352
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AFMID PubMed Links

Gene Details

Gene
AFMID
Gene Name
arylformamidase
There are no transcripts associated with this gene.

Gene
BIRC5
Gene Name
baculoviral IAP repeat containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012270.1 368 Intron NP_001012270.1
NM_001012271.1 368 Missense Mutation AGT,GGT S106G NP_001012271.1
NM_001168.2 368 Missense Mutation AGT,GGT S83G NP_001159.2

View Full Product Details