Product Details

SNP ID
rs200503858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8735423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACATTGCTGCGGGCACGGTCAAA[A/G]CGCATCTGCAGCCTTGCCAGCTCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC42 PubMed Links

Gene Details

Gene
CCDC42
Gene Name
coiled-coil domain containing 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001158261.1 908 Intron NP_001151733.1
NM_144681.2 908 Silent Mutation CGC,CGT R227R NP_653282.2

View Full Product Details