Product Details

SNP ID
rs199581006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28880594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACACCAGTGGCCTTCTTGATCAG[A/G]GGTATCTGCAAGGATGGGAGATGCC
Phenotype
MIM: 131560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLOT2 PubMed Links

Gene Details

Gene
FLOT2
Gene Name
flotillin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004475.2 1311 Silent Mutation CCC,CCT P418P NP_004466.2
XM_005257950.3 1311 Silent Mutation CCC,CCT P418P XP_005258007.1
XM_005257953.2 1311 Silent Mutation CCC,CCT P341P XP_005258010.1
XM_017024394.1 1311 Silent Mutation CCC,CCT P369P XP_016879883.1
XM_017024395.1 1311 Silent Mutation CCC,CCT P369P XP_016879884.1
XM_017024396.1 1311 Silent Mutation CCC,CCT P369P XP_016879885.1

View Full Product Details