Product Details

SNP ID
rs200297929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64510258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCACTGGCCTTGGACTACTTGCA[G/T]GTCAGCTTGTCTCACATAACAGGTT
Phenotype
MIM: 180630
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP95 PubMed Links

Gene Details

Gene
CEP95
Gene Name
centrosomal protein 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316990.1 340 UTR 5 NP_001303919.1
NM_138363.2 340 Missense Mutation CAG,CAT Q78H NP_612372.1
XM_005257779.3 340 Missense Mutation CAG,CAT Q78H XP_005257836.1
XM_005257780.2 340 UTR 5 XP_005257837.1
XM_006722166.3 340 UTR 5 XP_006722229.1
XM_011525458.2 340 Missense Mutation CAG,CAT Q78H XP_011523760.1
XM_017025287.1 340 UTR 5 XP_016880776.1
XM_017025288.1 340 UTR 5 XP_016880777.1
Gene
DDX5
Gene Name
DEAD-box helicase 5
There are no transcripts associated with this gene.

Gene
MIR3064
Gene Name
microRNA 3064
There are no transcripts associated with this gene.

Gene
MIR5047
Gene Name
microRNA 5047
There are no transcripts associated with this gene.

View Full Product Details