Product Details

SNP ID
rs200168432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49969699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGTCCTGCCAGCAACCCGCGGC[G/T]CTCTCTCAGCCCCTCTGCGGACCTG
Phenotype
MIM: 601911
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DLX4 PubMed Links

Gene Details

Gene
DLX4
Gene Name
distal-less homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001934.3 155 Intron NP_001925.2
NM_138281.2 155 Silent Mutation GCG,GCT A77A NP_612138.1
XM_017024291.1 155 UTR 5 XP_016879780.1

View Full Product Details