Product Details
- SNP ID
-
rs201985313
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:58305065 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGCCCAGCTCCTCACTGACCTGA[C/T]TCTCAGCCTGGGGTGTCCTGGGTTC
- Phenotype
-
MIM: 610764
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSPOAP1
PubMed Links
Gene Details
- Gene
- TSPOAP1
- Gene Name
- TSPO associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001261835.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1838S |
NP_001248764.1 |
NM_004758.3 |
5395 |
Missense Mutation |
AAT,AGT |
N1847S |
NP_004749.2 |
NM_024418.2 |
5395 |
Missense Mutation |
AAT,AGT |
N1787S |
NP_077729.1 |
XM_005257807.2 |
5395 |
Missense Mutation |
AAT,AGT |
N1838S |
XP_005257864.1 |
XM_006722173.2 |
5395 |
Missense Mutation |
AAT,AGT |
N1847S |
XP_006722236.1 |
XM_006722174.2 |
5395 |
Missense Mutation |
AAT,AGT |
N1840S |
XP_006722237.1 |
XM_006722175.2 |
5395 |
Missense Mutation |
AAT,AGT |
N1787S |
XP_006722238.1 |
XM_006722176.2 |
5395 |
Missense Mutation |
AAT,AGT |
N1570S |
XP_006722239.1 |
XM_006722177.3 |
5395 |
Missense Mutation |
AAT,AGT |
N1407S |
XP_006722240.1 |
XM_006722178.3 |
5395 |
Missense Mutation |
AAT,AGT |
N1407S |
XP_006722241.1 |
XM_011525471.2 |
5395 |
Missense Mutation |
AAT,AGT |
N1368S |
XP_011523773.1 |
XM_011525472.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1351S |
XP_011523774.1 |
XM_017025326.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1847S |
XP_016880815.1 |
XM_017025327.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1808S |
XP_016880816.1 |
XM_017025328.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1744S |
XP_016880817.1 |
XM_017025329.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1567S |
XP_016880818.1 |
XM_017025330.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1567S |
XP_016880819.1 |
XM_017025331.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1558S |
XP_016880820.1 |
XM_017025332.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1507S |
XP_016880821.1 |
XM_017025333.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1281S |
XP_016880822.1 |
XM_017025334.1 |
5395 |
Missense Mutation |
AAT,AGT |
N1230S |
XP_016880823.1 |
View Full Product Details