Product Details

SNP ID
rs201027671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3397481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTTATCATCAGAGAGAGAAGAA[A/G]GTTTTCTTCTGATGAATGACTCTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1E1 PubMed Links

Gene Details

Gene
OR1E1
Gene Name
olfactory receptor family 1 subfamily E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003553.2 930 Silent Mutation ACC,ACT T310T NP_003544.2

View Full Product Details