Product Details
- SNP ID
-
rs201577547
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:74357603 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGCCCACCGCCGGGCCCGCGCCT[C/T]CCGCCAGGTGCGCGCGCATGTAGTC
- Phenotype
-
MIM: 609046
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BTBD17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11547807] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BTBD17
- Gene Name
- BTB domain containing 17
- Gene
- GPR142
- Gene Name
- G protein-coupled receptor 142
There are no transcripts associated with this gene.
- Gene
- KIF19
- Gene Name
- kinesin family member 19
There are no transcripts associated with this gene.
View Full Product Details