Product Details

SNP ID
rs201780947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79834140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGCCGCCGCCACCGCCACCGCC[A/G]CCGACGCCGGTGTCTCGGGCTTCAC
Phenotype
MIM: 603079
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBX4 PubMed Links

Gene Details

Gene
CBX4
Gene Name
chromobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003655.2 3174 Missense Mutation GCG,GTG A501V NP_003646.2
XM_011525399.2 3174 Missense Mutation GCG,GTG A435V XP_011523701.1

View Full Product Details