Product Details

SNP ID
hCV190336337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42023671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCCAGGTCACCATCGTGGTCCT[C/T]GGCAACAAGTGTGACTTACAGGAGC
Phenotype
MIM: 601964 MIM: 604497
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC7 PubMed Links

Gene Details

Gene
DNAJC7
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
There are no transcripts associated with this gene.

Gene
NKIRAS2
Gene Name
NFKB inhibitor interacting Ras like 2
There are no transcripts associated with this gene.

Gene
ZNF385C
Gene Name
zinc finger protein 385C
There are no transcripts associated with this gene.

View Full Product Details