Product Details

SNP ID
rs201203253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60426197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAAGACCGCTGAAGAAGTTCCTG[C/T]GAAAGTTGCACCTGCCCAGGGACCT
Phenotype
MIM: 607740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf64 PubMed Links

Gene Details

Gene
C17orf64
Gene Name
chromosome 17 open reading frame 64
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181707.2 575 Nonsense Mutation CGA,TGA R64* NP_859058.2
XM_005257033.4 575 Nonsense Mutation CGA,TGA R97* XP_005257090.1
XM_005257034.4 575 Nonsense Mutation CGA,TGA R97* XP_005257091.1
XM_005257035.4 575 Nonsense Mutation CGA,TGA R27* XP_005257092.1
XM_005257036.2 575 Nonsense Mutation CGA,TGA R64* XP_005257093.1
XM_017024166.1 575 Nonsense Mutation CGA,TGA R27* XP_016879655.1
Gene
RPL12P38
Gene Name
ribosomal protein L12 pseudogene 38
There are no transcripts associated with this gene.

Gene
USP32
Gene Name
ubiquitin specific peptidase 32
There are no transcripts associated with this gene.

View Full Product Details