Product Details

SNP ID
rs201375481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75261913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCCAGCCAAGATGGCTGCCCCC[G/T]CAGTGAAGGTTGCCCGAGGATGGTC
Phenotype
MIM: 606006 MIM: 612072 MIM: 611974
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GGA3 PubMed Links

Gene Details

Gene
GGA3
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172703.2 246 Intron NP_001166174.1
NM_001172704.2 246 Intron NP_001166175.1
NM_001291641.1 246 Intron NP_001278570.1
NM_001291642.1 246 Intron NP_001278571.1
NM_014001.4 246 Intron NP_054720.1
NM_138619.3 246 Intron NP_619525.1
XM_011524563.2 246 Intron XP_011522865.1
XM_017024385.1 246 Intron XP_016879874.1
XM_017024386.1 246 Intron XP_016879875.1
XM_017024387.1 246 Intron XP_016879876.1
XM_017024388.1 246 Intron XP_016879877.1
XM_017024389.1 246 Intron XP_016879878.1
XM_017024390.1 246 Intron XP_016879879.1
XM_017024391.1 246 Intron XP_016879880.1
XM_017024392.1 246 Intron XP_016879881.1
Gene
LOC100287042
Gene Name
uncharacterized LOC100287042
There are no transcripts associated with this gene.

Gene
MIF4GD
Gene Name
MIF4G domain containing
There are no transcripts associated with this gene.

Gene
MRPS7
Gene Name
mitochondrial ribosomal protein S7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015971.3 246 Missense Mutation GCA,TCA A5S NP_057055.2

View Full Product Details