Product Details

SNP ID
rs200590185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49798415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGACATAGACATCTCCAGCCCCA[A/G]TGTATCTCACGATGAGAGCATTGCC
Phenotype
MIM: 609880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KAT7 PubMed Links

Gene Details

Gene
KAT7
Gene Name
lysine acetyltransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199155.1 653 Missense Mutation AAT,AGT N146S NP_001186084.1
NM_001199156.1 653 Intron NP_001186085.1
NM_001199157.1 653 Intron NP_001186086.1
NM_001199158.1 653 Intron NP_001186087.1
NM_007067.4 653 Missense Mutation AAT,AGT N146S NP_008998.1
XM_011524235.2 653 Intron XP_011522537.1

View Full Product Details