Product Details

SNP ID
rs201961696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5210842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTATTTGCAGGGATTTCAACAT[C/T]GTCATAGTCATCTTCAGGCTCAATG
Phenotype
MIM: 614406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100130950 PubMed Links

Gene Details

Gene
LOC100130950
Gene Name
uncharacterized LOC100130950
There are no transcripts associated with this gene.

Gene
SCIMP
Gene Name
SLP adaptor and CSK interacting membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271842.1 502 Missense Mutation AAT,GAT N126D NP_001258771.1
NM_001319190.1 502 Intron NP_001306119.1
NM_207103.3 502 Missense Mutation AAT,GAT N133D NP_996986.1
XM_017024615.1 502 Intron XP_016880104.1

View Full Product Details