Product Details

SNP ID
rs200763584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82019898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGAAGTGCAGGTGCAGCAGCCT[A/G]CTCACCAGCTCCTAGAAGGGAGGGG
Phenotype
MIM: 606236 MIM: 615128
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASPSCR1 PubMed Links

Gene Details

Gene
ASPSCR1
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251888.1 96 Intron NP_001238817.1
NM_024083.3 96 Intron NP_076988.1
XM_011523601.2 96 Intron XP_011521903.1
XM_011523602.2 96 Intron XP_011521904.1
XM_017025036.1 96 Intron XP_016880525.1
XM_017025037.1 96 Intron XP_016880526.1
XM_017025038.1 96 Intron XP_016880527.1
XM_017025039.1 96 Intron XP_016880528.1
XM_017025040.1 96 Intron XP_016880529.1
XM_017025041.1 96 Intron XP_016880530.1
XM_017025042.1 96 Intron XP_016880531.1
XM_017025043.1 96 Intron XP_016880532.1
XM_017025044.1 96 Intron XP_016880533.1
Gene
LRRC45
Gene Name
leucine rich repeat containing 45
There are no transcripts associated with this gene.

Gene
STRA13
Gene Name
stimulated by retinoic acid 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271006.1 96 Silent Mutation AGC,AGT S16S NP_001257935.1
NM_001271007.1 96 Silent Mutation AGC,AGT S16S NP_001257936.1
NM_144998.3 96 Silent Mutation AGC,AGT S16S NP_659435.2
XM_005256339.2 96 Silent Mutation AGC,AGT S16S XP_005256396.1
XM_017024326.1 96 Silent Mutation AGC,AGT S16S XP_016879815.1
XM_017024327.1 96 Silent Mutation AGC,AGT S16S XP_016879816.1
XM_017024328.1 96 Silent Mutation AGC,AGT S16S XP_016879817.1
XM_017024329.1 96 Silent Mutation AGC,AGT S16S XP_016879818.1

View Full Product Details