Product Details

SNP ID
rs201177829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74935817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAAATCGATGGGCCGCGGAGCC[C/T]GAGCGGCGGCTGCGCAGTCCCGCTG
Phenotype
MIM: 607827 MIM: 607828
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTOP2 PubMed Links

Gene Details

Gene
OTOP2
Gene Name
otopetrin 2
There are no transcripts associated with this gene.

Gene
OTOP3
Gene Name
otopetrin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272005.1 16 UTR 5 NP_001258934.1
NM_178233.2 16 Nonsense Mutation CGA,TGA R6* NP_839947.1
XM_011524744.2 16 Intron XP_011523046.1

View Full Product Details