Product Details

SNP ID
rs201625874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82316205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTCACACTGACCTGTGTCCTC[A/G]CCAGCACACACGCCACCCCCAGGCC
Phenotype
MIM: 186820 MIM: 602602
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD7 PubMed Links

Gene Details

Gene
CD7
Gene Name
CD7 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006137.6 711 Missense Mutation GCG,GTG A201V NP_006128.1
XM_011523608.1 711 Missense Mutation GCG,GTG A101V XP_011521910.1
XM_017025316.1 711 Missense Mutation GCG,GTG A101V XP_016880805.1
Gene
SECTM1
Gene Name
secreted and transmembrane 1
There are no transcripts associated with this gene.

View Full Product Details