Product Details

SNP ID
rs200976425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74357185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGCGCTGCCGTTGACGTCGAA[A/G]AACTTGGCGTAGTGCAGCGGCGACG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTBD17 PubMed Links

Gene Details

Gene
BTBD17
Gene Name
BTB domain containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080466.1 1751 Silent Mutation NP_001073935.1
XM_011524791.2 1751 Silent Mutation XP_011523093.1
XM_017024622.1 1751 Silent Mutation XP_016880111.1
Gene
KIF19
Gene Name
kinesin family member 19
There are no transcripts associated with this gene.

View Full Product Details