Product Details
- SNP ID
-
rs201045149
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:58305103 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTCCTGGGTTCCCTGTCCAGGCC[A/G]CCTGCCTCAGGCCCAGGGCCCTCCA
- Phenotype
-
MIM: 610764
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSPOAP1
PubMed Links
Gene Details
- Gene
- TSPOAP1
- Gene Name
- TSPO associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001261835.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1825G |
NP_001248764.1 |
NM_004758.3 |
5357 |
Silent Mutation |
GGC,GGT |
G1834G |
NP_004749.2 |
NM_024418.2 |
5357 |
Silent Mutation |
GGC,GGT |
G1774G |
NP_077729.1 |
XM_005257807.2 |
5357 |
Silent Mutation |
GGC,GGT |
G1825G |
XP_005257864.1 |
XM_006722173.2 |
5357 |
Silent Mutation |
GGC,GGT |
G1834G |
XP_006722236.1 |
XM_006722174.2 |
5357 |
Silent Mutation |
GGC,GGT |
G1827G |
XP_006722237.1 |
XM_006722175.2 |
5357 |
Silent Mutation |
GGC,GGT |
G1774G |
XP_006722238.1 |
XM_006722176.2 |
5357 |
Silent Mutation |
GGC,GGT |
G1557G |
XP_006722239.1 |
XM_006722177.3 |
5357 |
Silent Mutation |
GGC,GGT |
G1394G |
XP_006722240.1 |
XM_006722178.3 |
5357 |
Silent Mutation |
GGC,GGT |
G1394G |
XP_006722241.1 |
XM_011525471.2 |
5357 |
Silent Mutation |
GGC,GGT |
G1355G |
XP_011523773.1 |
XM_011525472.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1338G |
XP_011523774.1 |
XM_017025326.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1834G |
XP_016880815.1 |
XM_017025327.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1795G |
XP_016880816.1 |
XM_017025328.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1731G |
XP_016880817.1 |
XM_017025329.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1554G |
XP_016880818.1 |
XM_017025330.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1554G |
XP_016880819.1 |
XM_017025331.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1545G |
XP_016880820.1 |
XM_017025332.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1494G |
XP_016880821.1 |
XM_017025333.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1268G |
XP_016880822.1 |
XM_017025334.1 |
5357 |
Silent Mutation |
GGC,GGT |
G1217G |
XP_016880823.1 |
View Full Product Details