Product Details

SNP ID
rs201831800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16551863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGTTTCCCTTATCCATTAATTA[C/T]GATGTTTGGCACCTGTATGAACACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF287 PubMed Links

Gene Details

Gene
ZNF287
Gene Name
zinc finger protein 287
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020653.2 2732 Missense Mutation CAT,CGT H760R NP_065704.2
XM_011523968.2 2732 Missense Mutation CAT,CGT H760R XP_011522270.1
XM_017024887.1 2732 Missense Mutation CAT,CGT H760R XP_016880376.1

View Full Product Details