Product Details

SNP ID
rs202006891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43213834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTTCTCTCTCTCTAGAGCTGG[A/G]GAAGCAGTTGGTGGCTCTCATTCCC
Phenotype
MIM: 166945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00854 PubMed Links

Gene Details

Gene
LINC00854
Gene Name
long intergenic non-protein coding RNA 854
There are no transcripts associated with this gene.

Gene
NBR1
Gene Name
NBR1, autophagy cargo receptor
There are no transcripts associated with this gene.

Gene
TMEM106A
Gene Name
transmembrane protein 106A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291586.1 503 Missense Mutation GAG,GGG E73G NP_001278515.1
NM_001291587.1 503 Missense Mutation GAG,GGG E25G NP_001278516.1
NM_001291588.1 503 Missense Mutation GAG,GGG E73G NP_001278517.1
NM_145041.3 503 Missense Mutation GAG,GGG E73G NP_659478.1
XM_006721658.2 503 Missense Mutation GAG,GGG E73G XP_006721721.1
XM_006721659.3 503 Missense Mutation GAG,GGG E25G XP_006721722.1
XM_011524260.1 503 Missense Mutation GAG,GGG E73G XP_011522562.1
XM_017024113.1 503 Missense Mutation GAG,GGG E73G XP_016879602.1
XM_017024114.1 503 Missense Mutation GAG,GGG E25G XP_016879603.1

View Full Product Details