Product Details

SNP ID
rs202218976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34983206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGGAAGCCATCTAAGATCACG[C/T]GCCACCTACCTTGTTTTCTTGCCAG
Phenotype
MIM: 610730 MIM: 600940
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCT6B PubMed Links

Gene Details

Gene
CCT6B
Gene Name
chaperonin containing TCP1 subunit 6B
There are no transcripts associated with this gene.

Gene
LIG3
Gene Name
DNA ligase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002311.4 330 Silent Mutation CGC,CGT R67R NP_002302.2
NM_013975.3 330 Silent Mutation CGC,CGT R67R NP_039269.2
XM_005257970.3 330 Silent Mutation CGC,CGT R76R XP_005258027.1
XM_006721896.3 330 Silent Mutation CGC,CGT R76R XP_006721959.1
XM_017024624.1 330 Silent Mutation CGC,CGT R67R XP_016880113.1

View Full Product Details