Product Details

SNP ID
rs201536677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42980605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCTGTCGAAGCGGCTGCAGAGC[C/T]GGTAACGGTGGTGGCGGCTGTTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGES3L PubMed Links

Gene Details

Gene
PTGES3L
Gene Name
prostaglandin E synthase 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142653.1 44 Intron NP_001136125.1
NM_001142654.1 44 Intron NP_001136126.1
NM_001261430.1 44 Intron NP_001248359.1
Gene
PTGES3L-AARSD1
Gene Name
PTGES3L-AARSD1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136042.2 44 Intron NP_001129514.2
NM_025267.3 44 Intron NP_079543.1
Gene
RUNDC1
Gene Name
RUN domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321381.1 44 Missense Mutation CCG,CTG P10L NP_001308310.1
NM_173079.3 44 Missense Mutation CCG,CTG P10L NP_775102.2
XM_005257078.3 44 Missense Mutation CCG,CTG P10L XP_005257135.1
XM_005257080.3 44 Missense Mutation CCG,CTG P10L XP_005257137.1

View Full Product Details