Product Details

SNP ID
rs201845249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82019882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGGTCTTGTCATCCTTGAAGT[A/G]CAGGTGCAGCAGCCTGCTCACCAGC
Phenotype
MIM: 606236 MIM: 615128
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASPSCR1 PubMed Links

Gene Details

Gene
ASPSCR1
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251888.1 112 Intron NP_001238817.1
NM_024083.3 112 Intron NP_076988.1
XM_011523601.2 112 Intron XP_011521903.1
XM_011523602.2 112 Intron XP_011521904.1
XM_017025036.1 112 Intron XP_016880525.1
XM_017025037.1 112 Intron XP_016880526.1
XM_017025038.1 112 Intron XP_016880527.1
XM_017025039.1 112 Intron XP_016880528.1
XM_017025040.1 112 Intron XP_016880529.1
XM_017025041.1 112 Intron XP_016880530.1
XM_017025042.1 112 Intron XP_016880531.1
XM_017025043.1 112 Intron XP_016880532.1
XM_017025044.1 112 Intron XP_016880533.1
Gene
LRRC45
Gene Name
leucine rich repeat containing 45
There are no transcripts associated with this gene.

Gene
STRA13
Gene Name
stimulated by retinoic acid 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271006.1 112 Missense Mutation CAC,TAC H22Y NP_001257935.1
NM_001271007.1 112 Missense Mutation CAC,TAC H22Y NP_001257936.1
NM_144998.3 112 Missense Mutation CAC,TAC H22Y NP_659435.2
XM_005256339.2 112 Missense Mutation CAC,TAC H22Y XP_005256396.1
XM_017024326.1 112 Missense Mutation CAC,TAC H22Y XP_016879815.1
XM_017024327.1 112 Missense Mutation CAC,TAC H22Y XP_016879816.1
XM_017024328.1 112 Missense Mutation CAC,TAC H22Y XP_016879817.1
XM_017024329.1 112 Missense Mutation CAC,TAC H22Y XP_016879818.1

View Full Product Details