Product Details

SNP ID
rs199779499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:50227385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATATATTGATGCTCACGGAACTAC[G/T]GTGGAAGCTTACTGGGGCATGCCTT
Phenotype
MIM: 614759
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CFAP53 PubMed Links

Gene Details

Gene
CFAP53
Gene Name
cilia and flagella associated protein 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145020.4 1659 Missense Mutation NP_659457.2

View Full Product Details