Product Details
- SNP ID
-
rs200126026
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35033699 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTCGAAAACTACGAGCACAACAC[C/T]AGCGTCGTCAAGAAGATCCACATTG
- Phenotype
-
MIM: 142440
MIM: 600235
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GRAMD1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72558029] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GRAMD1A
- Gene Name
- GRAM domain containing 1A
There are no transcripts associated with this gene.
- Gene
- HPN
- Gene Name
- hepsin
There are no transcripts associated with this gene.
- Gene
- SCN1B
- Gene Name
- sodium voltage-gated channel beta subunit 1
View Full Product Details