Product Details

SNP ID
rs200233121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5785728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGATGTCCAGGCGCTCGGACGA[C/T]GAGATGTCCCAGTGCCGCTGCTCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CATSPERD PubMed Links

Gene Details

Gene
CATSPERD
Gene Name
cation channel sperm associated auxiliary subunit delta
There are no transcripts associated with this gene.

Gene
DUS3L
Gene Name
dihydrouridine synthase 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161619.1 1723 Silent Mutation TCA,TCG S300S NP_001155091.1
NM_020175.2 1723 Silent Mutation TCA,TCG S542S NP_064560.2
XM_017027020.1 1723 Silent Mutation TCA,TCG S528S XP_016882509.1
Gene
PRR22
Gene Name
proline rich 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134316.1 1723 Intron NP_001127788.1

View Full Product Details