Product Details

SNP ID
rs199503373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7730056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCAACGATCACCAGGTGCGCG[C/G]TGGCATCTGCGCAGTGATCCTGCGC
Phenotype
MIM: 604672 MIM: 616256
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD209 PubMed Links

Gene Details

Gene
CD209
Gene Name
CD209 molecule
There are no transcripts associated with this gene.

Gene
CLEC4G
Gene Name
C-type lectin domain family 4 member G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244856.1 659 Missense Mutation ACC,AGC T185S NP_001231785.1
NM_198492.3 659 Missense Mutation ACC,AGC T197S NP_940894.1

View Full Product Details